Hazard BinLiner™ Bags, Micronova
Supplier: Micronova
BL2-25REIRBB BL4-4060REPC
89236-646CS 643.2 CAD
89236-646 75779-994
Hazard BinLiner™ Bags, Micronova
Bags Waste Disposal Bags
These low density polyethylene Hazard BinLiner™ Bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
  • Low density polyethylene
  • Hazardous material bags are labeled for waste segregation in the U.S.
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Can be used with BLSSH series BinLiner™ Stands
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
The Biohazard liners are designed for containment and disposal of biomedicalMore Product Information