Micro-Touch® DermaClean® Latex Examination Gloves, Ansell
Supplier: Ansell Canada
3181-1 3180-1 3182-1
CA32892-800CS 262.94 CAD
CA32892-800 CA32892-798 CA32892-802
Micro-Touch® DermaClean® Latex Examination Gloves, Ansell
Gloves
These powder-free examination gloves have low protein content.
  • Textured outer surface provides an excellent grip in wet and dry conditions
  • Smooth inner surface for easy donning
  • Low allergenicity latex reduces risk of contact dermatitis due to absence of thiurams
This nonsterile glove has excellent grip levels making it ideal for healthcare providers who handle instrumentation, test tubes or infusion equipment. Ansell's natural rubber latex formulation ensures a comfortable, well-fitting glove while the smooth chlorinated inner surface facilitates easeMore Product Information