AlphaTec® 56-802 Endurosaf™ Aprons, Ansell
Supplier: Ansell Healthcare
951928 951929
75795-462CS 1310.55 CAD
75795-462 75795-464
AlphaTec® 56-802 Endurosaf™ Aprons, Ansell
Workwear
High-performance Endurosaf™ aprons feature a strong and durable 8-9 millimeter die-cut Enduro 2000™ polyurethane material that rivals traditional materials such as neoprene, nitrile, vinyl and other films.
  • Sealed ties provide a durable, consistent fit
  • Remain flexible in cold temperatures
  • Strong and tough fabrication
Sealed ties and adjustable neck straps are strong and provide a durable, consistent fit for a wide variety of users
Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact.More Product Information