AlphaTec® 87-298 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell
Supplier: Ansell Canada
0029811090
CA32890-631DZ 12.96 CAD
CA32890-631 CA32890-633 CA32890-635
AlphaTec® 87-298 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell
Gloves
Strength, sensitivity, moderate chemical protection and grip come together in a natural rubber latex glove that provides a high degree of tensile strength.
  • Excellent tactile sensitivity
  • Good protection against chemical splashes
  • Improved grip on wet or dry objects
The cotton flock lining helps maintain excellent sensitivity
Use in the automotive and transportation, chemical, food handling and processing, janitorial services, petrochemical, sanitation, and general industries. Uses include canning and food processing; chemical processing, oil refining and petrochemicals; handling acids, alkalies, salts, ketones and alcohols; and janitorial and maintenance work.
Gloves provide snag, puncture and abrasion resistance. The embossed pebble design provides improved grip on wet or dry objects.
Packaging: Gloves are packaged 12 pairs in a bag; 144 in a carton.