Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Supplier: Chemglass
CG-1981-10 CG-1981-12
80071-664EA 14.04 CAD
80071-664 80071-666
Spatulas, Rounded, Micro, Non-Magnetic, Chemglass
Spatulas
Spatulas are made of non-magnetic stainless steel and feature a mirror finish.
  • Spatula will pass through a 14/10 or 14/20 outer joint
  • Both ends are rounded and are approximately ⁵/₁₆" (8 mm) wide × 2" (51 mm) in length
  • Non-magnetic