Tuf Sponge Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
451.38X2X60BK 451.38X2X60BYL 451.38X2X3BK 451.38X2X3BYL 451.38X2X3GY 451.38X3X60BK 451.38X3X60BYL 451.38X3X60GY 451.38X3X5BK 451.38X3X12BK 451.38X3X5BYL 451.38X3X12BYL 451.38X3X5GY 451.38X3X12GY 451.38X4X60BK 451.38X4X60BYL 451.38X4X60GY 451.38X6X60BK
89020-686EA 94.15 CAD
89020-686 89020-688 89020-690 89020-692 89020-694 89020-696 89020-698 89020-700 89020-702 89020-704 89020-706 89020-708 89020-710 89020-712 89020-714 89020-716 89020-718 89020-720 89020-722 89020-724 89020-726 89020-728 89020-730 89020-732 89020-734
Tuf Sponge Anti-Fatigue Mat, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Tuf Sponge mats provide an economical PVC blown sponge barrier from hard floors.
  • Economic comfort for light industrial applications
  • Safety rounded edges
  • Can easily be cut to size on-site
Equipped with an attractive ribbed surface, this mat can be purchased in rolls and easily cut to length on-site for quick replacement when damaged. When the budget is tight, but leg and back fatigue must be battled, try Tuf Sponge. Ideally suited for use in dry environments.
Certifications: Meets ASTM F1677. Dry: COF = 0.94/Wet: COF = 0.72. Meets ASTM D575/0.205 @ 35 psi.
Order Now
SPECIFICATIONS
Environmentally Preferable