WeldSafe Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
447.916X4X75BK 447.916X2X75BK 447.916X2X3BK 447.916X3X5BK 447.916X3X75BK
89021-208EA 7902.91 CAD
89021-208 89021-210 89021-212 89021-214 89021-216
WeldSafe Anti-Fatigue Mat, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
WeldSafe mats feature a specially formulated SBR rubber surface that repels sparks and hot metal shards.
  • Thick rubber surface
  • Embossed leather texture
  • Melt point of over 500°F (260°C)
  • Unparalleled performance and comfort
The Nitrile enhanced Nitricell® sponge base absorbs repeated compression and bounces back to its original form better than any PVC sponge alternative. Accept no substitutes in critical welding applications. Ideally suited for use in dry environments.
Certifications: Meets ASTM F1677. Dry: COF = 1.10/Wet COF = 0.32. Meets ASTM D575.More Product Information