Cajon® Ultra-Torr Adapters, Chemglass
Supplier: Chemglass
AF-0066-09 AF-0066-10 AF-0066-11 AF-0066-12 AF-0066-13 AF-0066-14 AF-0066-16 AF-0066-17
80060-000EA 690.61 CAD
80060-000 80060-002 80060-004 80060-006 80060-008 80060-010 80060-012 80060-014
Cajon® Ultra-Torr Adapters, Chemglass
Tubing Transfer Tubing
Cajon® adapters provide the flexibility of rubber tubing.
  • Connections compensate for misalignment, expansion and contraction
  • Compressible by at least 20%
  • Extendable by 50% of its nominal listed length
Adapters are the ideal replacement for rubber, plastic, or glass tubing in critical vacuum applications.
Ordering information: Adapters are supplied complete with two AF-0067 unions, two AF-0068 cuff adapters and listed length of flexible tubing.