VWR® JerriCan
Supplier: VWR
VWR®
683225
75836-684CS 796.78 CAD
75836-684
VWR® JerriCan
Bottles Jerricans
These high-density polyethylene, disposable Jerri can are ideal for storing and transfer of Sterile fluids and pharmaceutical/biotech reagents.
  • Excellent chemical resistance
  • Leak-proof on standard test condition
  • Angled-neck opening is an advantage for pouring / transfering liquid
Manufactured with 100% virgin Medical grade resin meets US FDA and USP Class VI.
Carboys have excellent chemical resistance and are well suited for use from –100 to 100 °C. Graduated to identify the volume dispensed. Comes with integrated handle for easy carrying and pouring.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.