VWR® Round Carboy with Sanitary Flange, Narrow Mouth
Supplier: VWR International
75795-686 75795-694
75795-686EA 1099.78 CAD
75795-686 75795-694
VWR® Round Carboy with Sanitary Flange, Narrow Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile, excellent chemical resistance
  • Polypropylene formats available
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
  • Feature a 41 mm sanitary flange for use as a dispensing port
Carboys are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak-proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. Provides a secure hookup for use as a supply reservoir.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.