VWR® Round Carboy with Sanitary Neck
Supplier: VWR International
75795-678 75795-684 75795-692
75795-678CS 2211.63 CAD
75795-678 75795-684 75795-692
VWR® Round Carboy with Sanitary Neck
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for bioprocessing applications.
  • Non sterile
  • Polypropylene formats available
  • Excellent chemical resistant
  • Narrow-mouth
  • Feature an integrated shoulder hand
  • Features a 76 mm (3") sanitary flange that accepts standard TC fittings
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.