VWR® Rectangular Carboy, Wide Mouth
Supplier: VWR International
75795-672 75795-680 75795-696
75795-672CS 1077.62 CAD
75795-672 75795-680 75795-696
VWR® Rectangular Carboy, Wide Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene and HDPE formats available
  • Clear graduation marks
  • Excellent chemical resistance
  • Wide mouth
  • Heavy duty stainless steel handle for easy carrying and pouring
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.