Calibration Reference Materials for Autosamplers, DSC, PerkinElmer
Supplier: PerkinElmer
03190033 03190034 03190033
CA97012-532EA 179.38 CAD
CA97012-532 97012-532 97012-270
Calibration Reference Materials for Autosamplers, DSC, PerkinElmer
Standards Alloys and Metal Ore Standards
These calibration reference materials are 99.999% pure
DSC requires elements of this purity so that the instrument can be accurately calibrated.