Microflex® N89 Disposable Gloves, Ansell
Supplier: Ansell
Microflex®
N893 N891 N894 N892 N891 N892 N894 N893
CA10160-952BX 36.5 CAD
10160-950 10160-954 CA10160-952 10160-952 CA10160-950 CA10160-954 CA10160-948 10160-948
Microflex® N89 Disposable Gloves, Ansell
Gloves
The long cuff and increased thickness provides an additional level of hand protection for the user while the nitrile material offers superior puncture resistance.
  • A textured finish allows for a good grip
  • Longer length and thicker for added protection
Ideally used in analytical testing/measurements, food processing and handling, dairy processing, crime scene evaluation, and protection from blood and other bodily fluids.
Packaging: 50 gloves per box; 10 boxes per carton.