Sapphire 96-Well PCR Microplates, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
652230 652230P 652240 652285 652201 652250 652260 652290 669285 652210
75800-002CS 5493.41 CAD
75800-002 75800-004 75830-544 75830-550 75830-560 75830-562 75830-564 75830-568 75830-570 76199-202
Sapphire 96-Well PCR Microplates, Polypropylene, Greiner Bio-One
Microplates PCR Plates
96-Well PCR Microplates are manufactured out of medical-grade polypropylene and are compatible with a wide range of thermal cyclers.
  • Polypropylene construction
  • Free of RNase, DNase, and human DNA
  • Endotoxin-free
  • Suitable for real-time/quantitative PCR
Microplates are thin-walled for efficient heat transfer during amplification. Packaging is labeled with product symbols for easier identification and warehousing.