Accutech® 91-225 Sterile Rubber Latex Gloves, Powder-Free, Ansell
Supplier: Microflex
119742 119743 119744 119745 119746 119747 119748
10818-406CS 1156.81 CAD
10818-406 10818-408 10818-410 10818-412 10818-414 10818-416 10818-418
Accutech® 91-225 Sterile Rubber Latex Gloves, Powder-Free, Ansell
Gloves
Comfortable cleanroom gloves with low allergenic content and smooth grip.
  • Inner silicone coating facilitates easy donning and double gloving
  • Thinner fingertips and bisque finish provide greater sensitivity, increased handling precision, and dexterity
  • Low allergenic content minimizes the risk of latex-associated allergies for the wearer
Certifications: EN 374, Cat. III, CE marked.
Packaging: One pair per poly wallet, sealed in a poly envelope; 10 pairs per sealed polybag; 5 sealed polybags per master polybag; 4 master polybags per lined carton; 200 pairs per carton.
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Powdered/Powder-free Powder-free
Cuff style Straight