Microchem® by AlphaTec™ 68-1800 Industrial Coveralls with Low Hazard Liquid Protection and Bound Seams, Ansell
Supplier: Ansell Canada
WH18-B-92-103-02 WH18-B-92-103-03 WH18-B-92-103-04 WH18-B-92-103-05 WH18-B-92-103-06 WH18-B-92-103-07 WH18-B-92-111-06 WH18-B-92-111-07 WH18-B-92-111-08 WH18-B-92-111-09 WH18-B-92-111-02 WH18-B-92-111-03 WH18-B-92-111-04 WH18-B-92-111-05
10836-142CS 517.09 CAD
10836-142 10836-144 10836-146 10836-148 10836-150 10836-152 10836-154 10836-156 10836-338 10836-346 10836-348 10836-354 10836-356 10836-472 10836-474 10836-476 10836-478 10836-480 10836-482
Microchem® by AlphaTec™ 68-1800 Industrial Coveralls with Low Hazard Liquid Protection and Bound Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 1800 coveralls provide the wearer with entry level, low hazard liquid and particulate protection.
  • Low hazard liquid spray, splash, and particle protection
  • Moisture vapor permeable for breathability to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Antistatic tested according to EN1149-5 and AATCC 76
These disposable latex coveralls with bound seams have a 2-way front zipper with resealable storm flap and elasticated wrists, waist, and ankles. Ideal for a wide range of general industrial applications like general maintenance, paint spraying, powder handling, and food industries.
Certifications: Tested according to EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6.