Microchem® by AlphaTec™ 68-1500 PLUS FR Flame Retardant Coveralls with Serged Seams, Ansell
Supplier: Ansell Healthcare
NR17-S-92-111-02 NR17-S-92-111-03 NR17-S-92-111-05 NR17-S-92-111-06 NR17-S-92-111-07 NR17-S-92-111-08 NR17-S-92-111-09
10836-282CS 438.4 CAD
10836-282 10836-284 10836-286 10836-288 10836-290 10836-292 10836-294 10836-296 10836-298 10836-300 10836-302 10836-304 10836-484 10836-486 10836-488 10836-490
Microchem® by AlphaTec™ 68-1500 PLUS FR Flame Retardant Coveralls with Serged Seams, Ansell
Disposable Garments
MICROCHEM® by AlphaTec™ 1500 PLUS FR coveralls are highly breathable, flame retardant, and antistatic
  • Flame retardant
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Optimized body fit when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above)
  • Antistatic tested according to EN1149-5
Must be worn over thermal protective garments, such as NOMEX®, and never to be worn next to the skin. Coveralls can be used in the petrochemical industry and for industrial cleaning, utilities, and general maintenance.
Made of SMMS polypropylene nonwoven fabric, coveralls are designed for protection from particulates and light, non-flammable liquid spray or splash.
Certifications: Tested according to EN 1073-2, EN ISO 14116 / INDEX 1/0/0, EN 1149-5 / AATCC 76, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6, ANSI/ISEA 101.