Standard Corrugated Vinyl Runner, Wearwell®
Supplier: Wearwell
381.18X2X150BK 381.18X2X105BK 381.18X3X150BK 381.18X3X105BK 381.18X4X150BK 381.18X4X105BK
89021-278RL 1698.03 CAD
89021-278 89021-280 89021-282 89021-284 89021-286 89021-288
Standard Corrugated Vinyl Runner, Wearwell®
Floor Mats Permanent Floor Mats
Standard Corrugated Vinyl protects floors, reduces cleaning costs, and adds traction where it's needed.
  • Protects floors
  • Easy to clean, chemical resistant surface
  • Vinyl compound won't curl during use
Ideally suited for use in dry environments.
Certifications: Mats meet ASTM F1677. Dry: COF = 0.91/Wet: COF = 0.65.