Anchor Blade Impellers, Caframo
Supplier: Caframo
U022 U044 U055 U510 U044SW U044SW
CA58960-206EA 344.56 CAD
CA58960-206 CA27800-369 CA27800-370 CA27800-371 52428-760 CA52428-760
Anchor Blade Impellers, Caframo
Stirrers Stirring Tools
Caframo U-Shaped Anchor paddles are designed for viscous mixing.
  • Composed of electo-polished 316L stainless steel
  • Welded to the shaft for stability
  • Tangential flow
It is also referred to as a sweep, the blade offers high shear at the edges and can be used close to the vessel wall.