Bath Oils, Constant Temperature Bath, Cannon Instrument
Supplier: CANNON INSTRUMENT COMPANY CA
9726-L37 9726-L38 9726-L39 9726-L40 9726-L20 9726-L30 9726-L33 9726-L36
CA10021-110EA 1457.91 CAD
CA10021-110 CA10021-112 CA10021-114 CA10021-116 CA10022-010 CA10022-014 CA10022-016 CA10022-018
Bath Oils, Constant Temperature Bath, Cannon Instrument
Circulator Bath Fluids
Silicone bath oils are clear, colorless liquids that can be used as heat transfer media in Cannon automated viscometers and some high-temperature Cannon baths where IBF fluid is not effective.
  • Viscosity is 36 cSt at 40 °C, 5.6 cSt at 100 °C
  • Use below 135 °C (275 °F)
IBF is an ideal bath oil for Cannon heated baths with applications at ambient temperature and above. IBF Bath Oil contains an oxidation inhibitor which reduces tendency to darken at higher temperatures.
Order Now
SPECIFICATIONS
Environmentally Preferable