Difco™ Maltose (+), Monohydrate, BD Diagnostics
Supplier: BD Diagnostics
Difco™
216830 216830
CA90000-932EA 256.99 CAD
CA90000-932 90000-932
Difco™ Maltose (+), Monohydrate, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Maltose is a component of starch and glycogen, and is used as a nutrient in culture media.
  • Can be used as a sweetener
  • Mammalian and insect cell culture tested
Maltose can be further hydrolyzed to glucose by the action of maltase, an enzyme commonly found in yeast and many other sources.