BBL™ Mueller Hinton Broth, BD Life Sciences
Supplier: BD Life Sciences
BBL™
211443 296164 296164 211443
CA90000-194EA 223.72 CAD
90004-658 CA90000-194 CA-90004-658 90000-194
BBL™ Mueller Hinton Broth, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Mueller Hinton Broth is a general-purpose medium that may be used in the cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • Casein and beef extract provide essential nitrogen and nutrients
  • Starch protects against toxic substances that may be present
  • Dextrose produced by hydrolysis is a source of energy
Mueller Hinton Broth, unadjusted, has a formula similar to that of the solid medium, but without agar, for use when fluid medium is preferred. It may be used for the general cultivation of bacteria.