Polyethylene glycol monomethyl ether, M.W. 4000
Supplier: TCI America
P2187-100G
P2187-500G
TCP2187-100GEA
50.56
CAD
TCP2187-100G
TCP2187-500G
Polyethylene glycol monomethyl ether, M.W. 4000
Polyethylene glycol monomethyl ether
CAS Number: 9004-74-4
MDL Number: MFCD00084416
Form: Crystal
Color: White
Melting point (°C): 61
Flash Point (°C): 260
Other Product Names / Synonyms: mPEG
Formula:
CH₃(OCH₂CH₂)nOH Melting Pt: 61 °C Flash Pt: 260 °C |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...