Polyethylene glycol monomethyl ether, M.W. 2000

Supplier: TCI America

P2186-100G P2186-500G
TCP2186-100GEA 45.09 CAD
TCP2186-100G TCP2186-500G
Polyethylene glycol monomethyl ether, M.W. 2000
Polyethylene glycol monomethyl ether

CAS Number: 9004-74-4
MDL Number: MFCD00084416
Form: Crystal
Color: White
Melting point (°C): 53
Flash Point (°C): 254


Other Product Names / Synonyms: mPEG

Formula: CH₃(OCH₂CH₂)nOH
Melting Pt: 53 °C
Flash Pt: 254 °C
MDL Number: MFCD00084416
CAS Number: 9004-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89febb502f062dcb9eaa7ea049c611432bb27405&vid=89febb502f062dcb9eaa7ea049c611432bb27405&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR