3-Amino-4-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2301-5G
M2301-1G
TCM2301-5GEA
357.08
CAD
TCM2301-5G
TCM2301-1G
3-Amino-4-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)
3-Amino-4-methylbenzotrifluoride
CAS Number: 25449-96-1
MDL Number: MFCD00134580
Molecular Formula: C8H8F3N
Molecular Weight: 175.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 74
Melting point (°C): 40
Flash Point (°C): 96
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-4-methylbenzotrifluoride, 5-Trifluoromethyl-o-toluidine
Formula:
C₈H₈F₃N Boiling Pt: 74 °C Melting Pt: 40 °C Flash Pt: 96 °C |
MDL Number:
MFCD00134580 CAS Number: 25449-96-1 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...