N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0857-1G F0857-5G
TCF0857-1GEA 52.14 CAD
TCF0857-1G TCF0857-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine

CAS Number: 135673-97-1
MDL Number: MFCD00065614
Molecular Formula: C24H27NO4
Molecular Weight: 393.48
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 140
Specific rotation [a]20/D: -23 deg (C=1, DMF)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Fmoc-Cha-OH, N-Fmoc-3-cyclohexyl-L-alanine

Formula: C₂₄H₂₇NO₄
Melting Pt: 140 °C
MDL Number: MFCD00065614
CAS Number: 135673-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e49ffacbf49b4bfdf309c187c70fcdfe3f48922a&vid=e49ffacbf49b4bfdf309c187c70fcdfe3f48922a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR