Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0766-25G E0766-5G
TCE0766-25GEA 581.76 CAD
TCE0766-25G TCE0766-5G
Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 3-amino-1H-pyrazole-4-carboxylate

CAS Number: 6994-25-8
MDL Number: MFCD00005238
Molecular Formula: C6H9N3O2
Molecular Weight: 155.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: 3-Aminopyrazole-4-carboxylic Acid Ethyl Ester

Formula: C₆H₉N₃O₂
Melting Pt: 102 °C
MDL Number: MFCD00005238
CAS Number: 6994-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e74d0fb5882ab53c756a99e8a3406637d587c39&vid=8e74d0fb5882ab53c756a99e8a3406637d587c39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR