tert-Butyl-4-carbamoylpiperidine-1-carboxylate ≥97.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
tert-Butyl 4-carbamoylpiperidine-1-carboxylate, 1-Boc-4-piperidinecarboxamide, tert-Butyl 4-Carbamoyl-1-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxamide, 4-Carbamoyl-1-piperidinecarboxylic acid tert-Butyl ester
CAS Number: 91419-48-6
MDL Number: MFCD02180953
Molecular Formula: C11H20N2O3
Molecular Weight: 228.29
Purity/Analysis Method: >97.0% (GC,N)
Form: Crystal
Melting point (°C): 166
Other Product Names / Synonyms: 1-Boc-4-piperidinecarboxamide, tert-Butyl 4-Carbamoyl-1-piperidinecarboxylate, 4-Carbamoyl-1-piperidinecarboxylic Acid tert-Butyl Ester
Formula:
C₁₁H₂₀N₂O₃ MW: 228.2900 g/mol Melting Pt: 166 °C |
MDL Number:
MFCD02180953 CAS Number: 91419-48-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...