SP Bel-Art H-B DURAC® Bi-Metallic Dial Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61310-3400 B61310-5700
30617-788EA 37.78 CAD
30617-788 75841-286 75841-288 75841-294 75841-298 75841-300 75841-306 75841-308 75841-310 75841-312 75841-316 75841-320 75841-322 75842-704 75842-706 75842-710 75842-712 75842-714 75842-718 75852-862
SP Bel-Art H-B DURAC® Bi-Metallic Dial Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Minimum immersion is 50mm (2”) in liquids or 100mm (4”) in gases.
  • External adjustment nut allows for simple field calibration
Thermometers feature polycarbonate lens and probe cover with pocket clip (except 44mm (1.7”) and 50mm (2”) dial sizes).
Each thermometer features accuracy to 1%, stainless steel case and stem and is individually packaged with a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Test and Calibration.
The 44mm (1.7”) and 50mm (2”) dial sizes feature a beaker clip with built in adjustment tool in place of the probe cover with pocket clip.