Adapter, Addition Pipe for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1380-31 CLS-1380-40 CLS-1380-51
10170-318EA 377.95 CAD
10170-318 75780-224 75780-226
Adapter, Addition Pipe for Bioreactors, Chemglass
Bioreactors
This adapter can be used to add fluids or gases to the bio reactor
The portion of the adapter that is within the reactor has a short drip tip tube that can accept tubing to reach further into the listed capacity reactor. These Adapters are replacement parts for Applikon® Biotechnology and Broadley-James® brands of bioreactors.