Airfree Storage Flasks, Schlenk, Chemglass
Supplier: Chemglass
AF-0094-01 AF-0094-02 AF-0094-03 AF-0094-04 AF-0094-05
80064-334EA 388.5 CAD
80064-334 80064-336 80064-338 80064-340 80064-342
Airfree Storage Flasks, Schlenk, Chemglass
Flasks Round Bottom Flasks
Storage flask sidearm has a 9 mm OD tubulation for use with 3/8" Cajon® connections
Chem-Vac™ Chem-Cap® valve is used to control gas or liquid flow into the vessel.