MINUM-WARE® Conical Reaction Vials, Chemglass
Supplier: Chemglass
MW-80-04 MW-80-05
80064-210EA 77.6 CAD
80064-210 80064-212
MINUM-WARE® Conical Reaction Vials, Chemglass
Vials
Heavy wall vial has an internal conical bottom for optimum sample retrieval and flat base which provides added mechanical strength and bench stability.
  • Vials are graduated at specific intervals
  • With 14/10 threaded outer joint
Delivery information: Supplied complete with open-top screw cap, PTFE faced septa and Viton O-ring.