Portable Vacuum System, Chemglass
Supplier: Chemglass
AF-0300-01
80060-050EA 26299.37 CAD
80060-050
Portable Vacuum System, Chemglass
Pumps Vacuum Aspiration Systems
The Chemglass Airfree® portable vacuum system has been designed to be a totally self-contained system for use in vacuum distillations, sample degassing, vapor pressure and other preparative work requiring vacuum to 1 micron.