Hydrion™ Fractional Range pH Test Papers
Supplier: Micro Essential
315 345 390
470203-360CS 109.1 CAD
470203-360 470203-368 470203-372
Hydrion™ Fractional Range pH Test Papers
Test Papers pH Indicator Papers and Test Strips
Exhibit Sharp Color Change for Each 0.5 pH Interval
  • Supplied in single 15' rolls
  • Plastic dispenser and a color chart included