N-Methyl-N-(trimethylsilyl)trifluoroacetamide for GC derivatisation
Supplier: RESTEK CORP
|
Danger
|
Synonyms:
MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide
Silylation derivatization reagent.
- Replace active hydrogen, reducing polarity and making the compounds more volatile.
- Increase stability of derivatives.
Silylation is the most widely used derivatization procedure for sample analysis by GC. In silylation, an active hydrogen is replaced by an alkylsilyl group such as trimethylsilyl (TMS) or tert-butyldimethylsilyl (tert-BDMS). Silyl derivatives are more volatile, less polar, and more thermally stable. As a result, GC separation is improved and detection is enhanced. Both TMS and tert-BDMS reagents are suitable for a wide variety of compounds and can be used for many GC applications. (Note that silylation reagents are generally moisture-sensitive and must be sealed to prevent deactivation.)
Formula:
C₆H₁₂F₃NOSi MW: 199.25 g/mol Boiling Pt: 130 |
MDL Number:
MFCD00000411 CAS Number: 24589-78-4 EINECS: 246-331-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...