Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

34966-4X4L 34966-1L 34966-2.5L 34966-4X2.5L
BJ34966-4LCS 1175.43 CAD
BJ34966-4L BJ34966-1L BJ34966-2.5L BJ34966-4X2.5L
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Methanol

Methanol (CH3OH) is also known as Methyl Alcohol or wood alcohol. It is a clear, colorless, volatile and flammable liquid. Methanol is a common laboratory solvent and is especially useful for HPLC, UV/VIS spectroscopy, due to its low UV cutoff. Methanol is toxic if ingested and can result in blindness, coma and death. Suitable for LC-MS applications and spectrophotometry.


  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Formula: H₃COH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 1.11 (vs air)
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as HCOOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0005 %
Silver (Ag) Max. 0.1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Tin (Sn) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Absorbance at 210 nm Max. 0.523
Absorbance at 220 nm Max. 0.301
Absorbance at 230 nm Max. 0.125
Absorbance at 260 nm Max. 0.009
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
HPLC Gradient at 254 nm Max. 5 mAU
Suitability for the LC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR