Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

34860-4X4L 34860-2.5L 34860-2L 34860-4X2.5L 34860-4X2L
BJ34860-4LCS 1257.54 CAD
BJ34860-4L BJ34860-1L BJ34860-2.5L BJ34860-2L BJ34860-4X2.5L BJ34860-4X2L
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Methanol

Methanol (CH3OH) is also known as Methyl Alcohol or wood alcohol. It is a clear, colorless, volatile and flammable liquid. Methanol is a common laboratory solvent and is especially useful for chromatography due to its low UV cutoff. Methanol is toxic if ingested and can result in blindness, coma and death. Suitable for HPLC and spectrophotometry.


  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.03 %
Absorbance at 400 nm Max. 0.01
Absorbance 280 nm Max. 0.01
Absorbance at 260 nm Max. 0.04
Absobance at 254 nm Max. 0.02
Absorbance at 240 nm Max. 0.07
Absorbance at 235 nm Max. 0.10
Absorbance at 230 nm Max. 0.15
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 0.60
Absorbance at 205 nm Max. 1.00
Absorbance (JP)(Val.Ref) complying
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
APHA Max. 10
Vapor Pressure 410 mmHg ( 50 °C)
Vapor Density 1.11 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR