HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
34812-6X1L-US 34816-6X1L-US 34805-6X1L-US 34805-4X2.5L-US 34805-6X500ML-US 34806-6X1L-US 34806-6X500ML-US 34698-6X1L-US 34800-6X1L-US 34800-4X2.5L-US 34730-2.5L
BJ34698-1LCS 1266.26 CAD
BJ34698-1L BJ34800-1L BJ34801-1L BJ34806-1L BJ34811-1L BJ34812-1L BJ34816-1L BJ37817-1L BJ34817-1L BJ34805-1L-US BJ34805-2.5LUS BJ34805500MLUS BJ34806-1L-US BJ34806500MLUS BJ34698-1L-US BJ34800-1L-US BJ34800-2.5LUS BJ-34730-2.5L
HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
Karl Fischer Titrants and Reagents
Original and Innovative Karl Fischer Reagents to analyze or measure water content
Our premium Hydranal™ product range for pyridine-free and ethanol-based water determination by Karl Fischer titration (KF titration) is comprised of one- and two-component reagents for volumetric Karl Fischer titrations. Also available are water standards for titer standardization, pH buffering reagents, special reagents for aldehydes, ketones, and other solubilizing agents.