Microcentrifuge Tubes, Flat Top, Bio Plas
Supplier: Bio Plas
4300 4303 4304 4306 4310 4301 4302 4307 4315 4316 4317 4318 4319 4320 4322 4325
20170-841PK 78.38 CAD
20170-841 20170-847 20170-849 20170-853 20170-835 20170-843 20170-845 20170-851 20170-855 20170-857 20170-859 20170-861 20170-863 20170-865 20170-866 20170-870 20170-872
Microcentrifuge Tubes, Flat Top, Bio Plas
Tubes Microcentrifuge Tubes
The Bio Plas tube is built to withstand speeds over 22000 ×g
  • Graduated from 0.1 to 1.5 ml
  • Easy-to-open with thumb pressure
  • Non wetted wall
Flat top lid forms an extra-tight seal that will not come open during processing. It features textured writing surfaces on both the lid and sides.
Order Now
SPECIFICATIONS
Environmentally Preferable