60 MHz Benchtop NMR Spectrometers, Nanalysis
Supplier: NANALYSIS CORP. CA
N60-2009-C-0031 N60-2009-C N60-2009-C-0011 4444 N60-2009-C-0013 2525 N60-2009-C-0007
CA76543-028EA 68760.23 CAD
CA76543-028 CA76543-030 CA76543-032 CA76543-034 CA76543-038 CA76543-040 CA76543-042
60 MHz Benchtop NMR Spectrometers, Nanalysis
Spectrophotometers and Photometers NMR Spectrometers
The Nanalysis 60 MHz benchtop NMR Spectrometers are compact 60 MHz, 1.4 T, all-in-one spectrometers that are both easy-to-use and easy-to-site wherever you need it most e.g., benchtop, fumehood, glovebox.
  • 5 mm NMR tubes
  • Optional flow kit
  • 1D and 2D NMR experiments
  • Multinuclear detection
  • Low maintenance
  • Easy-to-use
The Nanalysis 60 MHz benchtop NMR Spectrometers are compact 1.4 T, all-in-one spectrometers (magnet, electronics and computer in a single enclosure) that are both easy-to-use and easy-to-site. With good performance and high sensitivity fromMore Product Information