Qualakote®EDSNY PDESDNY Polyurethane Palm Dip ESD Nylon Gloves, QRP
Supplier: PIP
PDESDNYS PDESDNYM PDESDNYL PDESDNYXL PDESDNY-2X
89040-974CS 1990.74 CAD
89040-974 89040-976 89040-978 89040-980 89040-982
Qualakote®EDSNY PDESDNY Polyurethane Palm Dip ESD Nylon Gloves, QRP
Gloves
The QRP PDESDNY Palm Dip ESD Nylon Assembly Gloves offers micro-foamed polyurethane palm coating for an excellent grip.
  • Nylon + conductive carbon/nylon yarn
  • Uncoated back, seamless knit, for breathability, comfort
  • Human Body Model ANSI/ESD SP15.1 CAFE` resistance: 10⁷ uncoated area; 10⁸¯⁹ coated area
  • Machine washable for economy of repeated use
RoHS Compliant, PDESDNY Nylon Assembly Glove is knit from textured nylon yarn and features a micro-foamed polyurethane dip palm for an excellent grip. Seamless knit for greater comfort.