Qualatrile®XC 9C Nitrile Latex-Free Fingercots, QRP
Supplier: PIP
9CS 9CM 9CL 9CXL
14003-420CS 3968.49 CAD
14003-420 14003-422 14003-424 32889-139
Qualatrile®XC 9C Nitrile Latex-Free Fingercots, QRP
Finger Cots Nitrile Finger Cots
The QRP 9C Nitrile Latex-Free Finger Cots have an average surface resistacne of 5×10⁹ ohms per square unit.
  • Packaged for ISO 5 (Class 100) applications
  • For use with Class II static sensitive devices (thresholds above 1000V)
  • Unique, stand-up pouch - easy product identification
Order Now
SPECIFICATIONS
Environmentally Preferable