Qualatex® XC 8J Static Dissipative Latex Fingercots, QRP
Supplier: PIP
8JS 8JM 8JL 8JXL
89066-042CS 2798.75 CAD
89066-042 89066-044 89066-046 89066-048
Qualatex® XC 8J Static Dissipative Latex Fingercots, QRP
Finger Cots Latex Finger Cots
The QRP 8J Static Dissipative Latex Finger Cots have an average surface resistance of 10 ohms per square surface unit.
  • Packaged for ISO 5 (Class 100) applications
  • Compliant with ASTM and IEST-RP-CC 005.3 standards
  • Average surface resistance of 10 ohms per square surface unit
  • Fore use with Class I and Class II static sensitive devices
The QRP 8C Static Dissipative Latex Finger Cots have an average surface resistance of 10⁷ ohms per square surface unit. Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Compliant with ASTM and IEST-RP-CC 005.3 standards. For use with Class II static sensitive devices (thresholds above 1000V). Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2.
Certifications: ESD Safe to ANSO/ESD S20.20-2014.