Qualatex® XC 8C Static Dissipative Latex Fingercots, QRP
Supplier: QRP
8CS 8CXL
32888-922CS 2986.82 CAD
32888-922 32888-916
Qualatex® XC 8C Static Dissipative Latex Fingercots, QRP
Finger Cots Latex Finger Cots
The QRP 8C Static Dissipative Latex Finger Cots have an average surface resistance of 10 ohms per square surface unit.
  • Packaged for ISO 5 (Class 100) applications
  • Compliant with ASTM and IEST-RP-CC 005.3 standards
  • Average surface resistance of 10 ohms per square surface unit
  • Fore use with Class I and Class II static sensitive devices
  • Unique, Stand-Up pouch - easy product identification
The QRP 8C Static Dissipative Latex Finger Cots have an average surface resistance of 10 ohms per square surface unit. Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Compliant with ASTM and IEST-RP-CC 005.3 standards. For use with Class II static sensitive devices (thresholds above 1000V). Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
Certifications: ESD Safe to ANSO/ESD S20.20-2014.