Qualatrile® 8BQF09 Qualatrile Blue Nitrile 9" Powder-Free Gloves, QRP
Supplier: PIP
8BQF09S 8BQF09M 8BQF09L 8BQF09XL
89066-026CS 845.57 CAD
89066-026 89066-028 89066-030 89066-032
Qualatrile® 8BQF09 Qualatrile Blue Nitrile 9" Powder-Free Gloves, QRP
Gloves
The QRP 8BQF09 Qualatrile Blue Nitrile 9" powder-free gloves are naturally anti-static with a surface resistance of 10⁹ ohms.
  • Rugged 8 mil thickness
  • 3× puncture resistance of vinyl or natural rubber latex gloves
  • Geogrip 360sm fully textured fingers and palms
RoHS Compliant, 9" Qualatrile Blue 8 mil Powder-Free nitrile ambidextrous gloves are the professional's choice for exceptional personnel, product and process protection
Beaded cuff for added strength and easier donning. Textured for enhanced wet/dry grip without compromising tactile sensitivity. Manufactured in FDA-registered facilities utilizing an FDA-registered 510(k) formulation. Meets US government 21-CFR specifications; acceptable for use in food and pharmaceutical applications.
Inherently anti-static Qualatrile Blue Nitrile features superior snag and puncture resistance plus excellent flexibility and tactile sensitivity, while protecting the wearer from a broad range of common chemicals.
Certifications: Meets ASTM D6319.