Qualatex® XC 7C Anti-Static Latex Fingercots, QRP
Supplier: PIP
7CS 7CM 7CL 7CXL
56614-007CS 2360.24 CAD
56614-007 56614-018 56614-029 56614-030
Qualatex® XC 7C Anti-Static Latex Fingercots, QRP
Finger Cots Latex Finger Cots
The QRP 7C Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices.
  • Packaged for ISO 5 (Class 100) applications
  • Compliant with ASTM and IES-RP-CC 005.3 standards
  • Average surface resistance of 5 X 10¹² ohms per square unit
  • For use with Class II static sensitive devices (thresholds above 1000V)
  • Unique, Stand-Up pouch - easy product identification
7C pink Anti-Static Clean Room Latex Finger Cots are recommended for use with static sensitive devices. The QRP 7C gloves are packaged for ISO 5 (Class 100) applications and are compliant with ASTM and IES-RP-CC 005.3 standards. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
Certifications: ESD Safe to ANSI/ESD S20.20-2014.
Caution: This product contains natural rubber latex which may cause allergic reactions.