Qualatherm® 70G Cleanroom Gloves, 450 Wet/Dry Thermal Protection, QRP
Supplier: PIP
70GL 70GM
32885-780PK 1486.65 CAD
32885-780 32889-013
Qualatherm® 70G Cleanroom Gloves, 450 Wet/Dry Thermal Protection, QRP
Gloves
The QRP 70G Qualatherm Cleanroom Mid-Temperature Gloves are RoHS compliant and have a PolyTuff silicon elastomer formulation that offers clean, liquid-proof mid-temperature protection for wet or dry environments.
  • Packaged for ISO 5 (Class 100)
  • Wet or dry contact temperatures from –78 to 450 °F (–61.4 to 232 °C)
  • PolyTuff silicone elastomer is sulfur-free
  • Excellent resistance to chemicals, solder, and fluxes
  • Free from PCB, asbestos and fiberglass hazards
  • Latex-free
  • Non-particulating, cut and abrasion resistant
Seamless Comfort Curve™ paired Right Hand/Left Hand gloves minimize hand and finger fatigue. Micro-coated, high density, continuous filament nylon liner assures clean, easy donning and removal.
Applications of the 70G Qualatherm 450 °F mid-temperature cleanroom gloves include: diffusion furnaces, Quartz wafer boats, burn-in ovens, PWB handling after wave soldering, ceramics, porcelains, food processing and handling.