Qualatex® HiProXC 612HC Qualatex HiPro XC Latex 12" AMBI Gloves, QRP
Supplier: PIP
612HC-2X 612HCXL 612HCL 612HCM 612HCS
14003-586CS 1355.88 CAD
14003-586 14003-588 14003-590 14003-592 14003-594
Qualatex® HiProXC 612HC Qualatex HiPro XC Latex 12" AMBI Gloves, QRP
Gloves
Premium natural rubber latex gloves for ISO 5 (Class 100) applications.
  • Lowest extractables and particulates
  • Free from plasticizers, silicone, and powders
612HC Qualatex HiPro XC Latex Ambi Clean Room Gloves are RoHS compliant, 6 mil, 12" length, AMBI, and have textured palm and fingers.
Applications: IC and disk drive manufacture, electronics, laboratories, precision optics, assembly.
Packed 100 gloves/PBIB pack, 10 packs/case. Hi-Pro series latex gloves are manufactured free of traditional silicone oil and mold release agents which can be a primary contaminant source.
Certifications: Meets or exceeds all applicable requirements of IEST-RP-CC005.2.
Caution: This product contains natural rubber latex which may cause allergic reactions.