Qualatex® XC 5J Powder-Free Latex Fingercots, QRP
Supplier: PIP
5JS 5JM 5JL 5JXL
89066-034CS 1345.62 CAD
89066-034 89066-036 89066-038 89066-040
Qualatex® XC 5J Powder-Free Latex Fingercots, QRP
Finger Cots Latex Finger Cots
Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers.
  • Packaged for ISO 5 (Class 100) applications
  • Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers
The 5J Qualatex® powder-free cleanroom latex finger cots are packaged for ISO 5 (Class 100) applications
Applications: Discrete components, hard drive assemblies, die bonding, wire attach, disk drives/thin film heads, qoptics, finishing, cleaning & handling of precision instruments.
Compliant with ASTM and IEST-RP-CC 005.3 standards. Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Finger cots are packaged in gas-impremeable film flushed with semiconductor grade N2. Unique, Stand-Up Pouch provides easy product identification.
Certifications: Compliant with ASTM and IEST-RP-CC 005.3 standards.
Caution: This product contains natural rubber latex which may cause allergic reactions.